2-Chloro-2'-methylacetanilide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-N-(o-tolyl)acetamide

A535989-250MG A535989-5G
77266-046EA 35.02 USD
77266-046 77266-048
2-Chloro-2'-methylacetanilide 95%
2-Chloro-2'-methylacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.6348 g/mol
MDL Number: MFCD00018894
CAS Number: 37394-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR