3,3'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propane-1,2-diol) 95%

Supplier: Ambeed
Warning

A535828-250MG A535828-100MG
77253-630EA 211 USD
77253-630 77572-694
3,3'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propane-1,2-diol) 95%
3,3'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propane-1,2-diol)

Precaución: for Research Use Only

Formula: C₂₁H₂₈O₆
MDL Number: MFCD01862948
CAS Number: 5581-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR