BRD9539 98%

Supplier: Ambeed
Warning

A535666-10MG A535666-1MG A535666-5MG
77266-040EA 87 USD
77266-040 77266-042 77266-044
BRD9539 98%
BRD9539

Precaución: for Research Use Only

Formula: C₂₄H₂₁N₃O₃
MDL Number: MFCD23704793
CAS Number: 1374601-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR