3-Iodobenzonitrile 98%

Supplier: Ambeed
Warning

A534765-10G A534765-1G A534765-25G A534765-500G A534765-5G
77279-898EA 25.75 USD
77279-898 77279-900 77279-902 77279-904 77279-906
3-Iodobenzonitrile 98%
3-Iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄IN
MW: 229.02 g/mol
MDL Number: MFCD00079762
CAS Number: 69113-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR