Ethyl 2-((2,6-dimethylphenyl)amino)-2-oxoacetate 97%

Supplier: Ambeed
Danger

A534031-100MG A534031-1G A534031-250MG A534031-5G
77266-032EA 5.15 USD
77266-032 77266-034 77266-036 77266-038
Ethyl 2-((2,6-dimethylphenyl)amino)-2-oxoacetate 97%
Ethyl 2-((2,6-dimethylphenyl)amino)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.25 g/mol
MDL Number: MFCD00735403
CAS Number: 2903-46-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR