DL-O-Tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(2-hydroxyphenyl)propanoic acid, H-DL-o-Tyr-OH, 2-Hydroxyphenylalanine, 3-(o-Hidroxifenil)-DL-alanina

A533616-1G A533616-250MG A533616-100MG A533616-5G
77253-608EA 140 USD
77253-608 77253-610 77747-088 77747-089
DL-O-Tyrosine 98%
DL-O-Tyrosine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD00021725
CAS Number: 2370-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR