2,6-Dibromo-3,5-difluoroisonicotinic acid 98%

Supplier: Ambeed
Danger

A533095-100MG A533095-250MG
77253-604EA 123.6 USD
77253-604 77253-606
2,6-Dibromo-3,5-difluoroisonicotinic acid 98%
2,6-Dibromo-3,5-difluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆HBr₂F₂NO₂
MW: 316.88 g/mol
MDL Number: MFCD03411655
CAS Number: 325461-60-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR