(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine
A533066-100MG
A533066-1G
A533066-250MG
A533066-5G
77241-802EA
66.95
USD
77241-802
77241-804
77241-806
77241-808
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine 98%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine
Precaución: for Research Use Only
Formula:
C₃₆H₅₆FeP₂ MW: 606.6300 g/mol |
MDL Number:
MFCD01074441 CAS Number: 167416-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...