5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)pentan-1-one 97%

Supplier: Ambeed
Warning

Synonyms: Tetrahydropiperine

A532731-250MG A532731-1G A532731-5G
77266-030EA 46.35 USD
77266-030 77594-642 77749-113
5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)pentan-1-one 97%
5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)pentan-1-one

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₃
MW: 289.36 g/mol
MDL Number: MFCD17167032
CAS Number: 23434-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR