4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoic acid 95%

Supplier: Ambeed
Warning

A532297-100MG A532297-1G A532297-250MG
77292-324EA 144.2 USD
77292-324 77292-326 77292-328
4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoic acid 95%
4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₅N₇O₂
MW: 325.32 g/mol
MDL Number: MFCD00075774
CAS Number: 19741-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR