4-Methoxybenzyl isothiocyanate 97%

Supplier: Ambeed
Danger

A532291-1G A532291-5G A532291-25G
77292-320EA 23 USD
77292-320 77292-322 77617-266
4-Methoxybenzyl isothiocyanate 97%
4-Methoxybenzyl isothiocyanate

Precaución: for Research Use Only

Formula: C₉H₉NOS
MDL Number: MFCD00041117
CAS Number: 3694-57-3
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR