(R)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 97%

Supplier: Ambeed
Warning

A532278-100MG A532278-1G A532278-250MG
77266-024EA 51 USD
77266-024 77266-026 77266-028
(R)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 97%
(R)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂O
MW: 211.08 g/mol
MDL Number: MFCD30749348
CAS Number: 2097958-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR