2-Phenylethyl propionate 98%

Supplier: Ambeed
Warning

Synonyms: Propionic Acid Phenethyl Ester, Propionic Acid 2-Phenylethyl Ester, 2-Phenylethyl Propionate, Phenethyl Propionate

A531725-1000G A531725-100G A531725-25G A531725-500G
77279-890EA 55.62 USD
77279-890 77279-892 77279-894 77279-896
2-Phenylethyl propionate 98%
2-Phenylethyl propionate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00027008
CAS Number: 122-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR