3-Phenylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Aminomethyl)biphenyl, [1,1'-Biphenyl]-3-ylmethanamine

A531443-5G A531443-1G A531443-10G A531443-250MG
77292-316EA 138 USD
77292-316 77569-030 77594-640 77749-109
3-Phenylbenzylamine 98%
3-Phenylbenzylamine

Precaución: For research use only.

Formula: C₁₃H₁₃N
MDL Number: MFCD00270124
CAS Number: 177976-49-7
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR