Silibinin 95%

Supplier: Ambeed
Warning

Synonyms: Silybin A, (2R,3R)-3,5,7-Trihydroxy-2-((2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one

A531148-1G A531148-5G
77253-596EA 13 USD
77253-596 77253-598
Silibinin 95%
Silibinin

Precaución: for Research Use Only

Formula: C₂₅H₂₂O₁₀
MW: 482.44 g/mol
MDL Number: MFCD03225424
CAS Number: 22888-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR