2,6-Dichloroisonicotinamide 95%

Supplier: Ambeed
Warning

A531049-10G A531049-25G
77253-592EA 127 USD
77253-592 77253-594
2,6-Dichloroisonicotinamide 95%
2,6-Dichloroisonicotinamide

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MDL Number: MFCD00052640
CAS Number: 89281-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR