2'-Deoxyguanosine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3,9-dihydro-6H-purin-6-one

A530913-100MG A530913-1MG A530913-25G A530913-5G A530913-100G A530913-1G
77279-876EA 6.18 USD
77279-876 77279-878 77279-880 77569-026 77617-262 77617-264
2'-Deoxyguanosine 95%
2'-Deoxyguanosine

Precaución: For research use only.

Formula: C₁₀H₁₃N₅O₄
MW: 267.24 g/mol
MDL Number: MFCD00080300
CAS Number: 961-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR