2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol 95%

Supplier: Ambeed
Warning

A530638-10G A530638-1G A530638-250MG A530638-25G A530638-5G A530638-100G
77253-582EA 128 USD
77253-582 77253-584 77253-586 77253-588 77253-590 77747-086
2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol 95%
2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
MDL Number: MFCD00185588
CAS Number: 2162-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR