2-Ethyl 5-methyl 1H-indole-2,5-dicarboxylate 95%

Supplier: Ambeed
Warning

A530183-100MG A530183-1G A530183-250MG
77279-870EA 16 USD
77279-870 77279-872 77279-874
2-Ethyl 5-methyl 1H-indole-2,5-dicarboxylate 95%
2-Ethyl 5-methyl 1H-indole-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD06656020
CAS Number: 884494-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR