2-Methyl-3-nitrobenzotrifluoride 98%

Supplier: Ambeed
Danger

A530162-10G A530162-25G A530162-5G
77241-790EA 23 USD
77241-790 77241-792 77241-794
2-Methyl-3-nitrobenzotrifluoride 98%
2-Methyl-3-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MDL Number: MFCD00042322
CAS Number: 6656-49-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR