3-((2-Methylallyl)thio)-5H-[1,2,4]triazino[5,6-b]indole 99+%

Supplier: Ambeed
Warning

A530038-10MG A530038-1MG A530038-25MG A530038-50MG A530038-5MG
77253-572EA 57 USD
77253-572 77253-574 77253-576 77253-578 77253-580
3-((2-Methylallyl)thio)-5H-[1,2,4]triazino[5,6-b]indole 99+%
3-((2-Methylallyl)thio)-5H-[1,2,4]triazino[5,6-b]indole

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₄S
MDL Number: MFCD02369534
CAS Number: 328023-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR