3-Amino-3-[2-(trifluoromethyl)phenyl]propionic Acid 95%

Supplier: Ambeed
Warning

A529721-100MG A529721-1G A529721-250MG
77279-864EA 29.87 USD
77279-864 77279-866 77279-868
3-Amino-3-[2-(trifluoromethyl)phenyl]propionic Acid 95%
3-Amino-3-[2-(trifluoromethyl)phenyl]propionic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
MDL Number: MFCD01250834
CAS Number: 299165-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR