N-(4-Hydroxyphenyl)-4-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A529591-1G A529591-250MG A529591-5G
77241-784EA 29 USD
77241-784 77241-786 77241-788
N-(4-Hydroxyphenyl)-4-methylbenzenesulfonamide 97%
N-(4-Hydroxyphenyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃S
MDL Number: MFCD00183142
CAS Number: 1146-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR