Bay K 8644 98%

Supplier: Ambeed
Warning

Synonyms: (+-)-Bay K 8644, (±)-Bay K 8644, Methyl 2,6-dimethyl-5-nitro-4-(2-(trifluoromethyl)phenyl)-1,4-dihydropyridine-3-carboxylate

A529478-10MG A529478-1MG A529478-25MG A529478-50MG A529478-5MG
77266-006EA 156.56 USD
77266-006 77266-008 77266-010 77266-012 77266-014
Bay K 8644 98%
Bay K 8644

Precaución: for Research Use Only

Formula: C₁₆H₁₅F₃N₂O₄
MW: 356.301 g/mol
MDL Number: MFCD00036697
CAS Number: 71145-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR