Ethyl 6-fluoro-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Ethoxycarbonyl)-6-fluoro-1H-indole

A529310-100MG A529310-1G A529310-250MG A529310-5G
77279-852EA 14.42 USD
77279-852 77279-854 77279-856 77749-101
Ethyl 6-fluoro-1H-indole-2-carboxylate 97%
Ethyl 6-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₂
MW: 207.201 g/mol
MDL Number: MFCD03095203
CAS Number: 348-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR