1-Bromohexadecane 95%

Supplier: Ambeed
Warning

A528920-1000G A528920-100G A528920-25G A528920-500G
77241-772EA 43.26 USD
77241-772 77241-774 77241-776 77241-778
1-Bromohexadecane 95%
1-Bromohexadecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₁₄CH₃
MW: 305.34 g/mol
MDL Number: MFCD00000230
CAS Number: 112-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR