Bis(2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)phenyl)amine 97%

Supplier: Ambeed
Warning

A528801-100MG A528801-250MG
77266-000EA 65 USD
77266-000 77266-002
Bis(2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)phenyl)amine 97%
Bis(2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)phenyl)amine

Precaución: for Research Use Only

Formula: C₃₀H₂₅N₃O₂
CAS Number: 485394-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR