(S)-1-{(S)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-ferrocenyl}ethyldi(3,5-xylyl)phosphine 98%

Supplier: Ambeed
Warning

A528733-100MG A528733-250MG A528733-1G
77253-568EA 36 USD
77253-568 77253-570 77617-252
(S)-1-{(S)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-ferrocenyl}ethyldi(3,5-xylyl)phosphine 98%
(S)-1-{(S)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-ferrocenyl}ethyldi(3,5-xylyl)phosphine

Precaución: for Research Use Only

Formula: C₄₄H₃₆F₁₂FeP₂
MDL Number: MFCD08561154
CAS Number: 1272317-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR