2-Nitro-p-cymene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene

A528568-10G A528568-1G A528568-25G A528568-5G
77279-844EA 94.76 USD
77279-844 77279-846 77279-848 77279-850
2-Nitro-p-cymene 97%
2-Nitro-p-cymene

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00007176
CAS Number: 943-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR