2,4,8-Trichloro-6-fluoroquinazoline 97%

Supplier: Ambeed
Warning

A528551-100MG A528551-1G
77265-996EA 285.31 USD
77265-996 77265-998
2,4,8-Trichloro-6-fluoroquinazoline 97%
2,4,8-Trichloro-6-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₂Cl₃FN₂
MW: 251.47 g/mol
CAS Number: 1702815-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR