5,9-Dibromo-7H-benzo[c]fluoren-7-one 98+%

Supplier: Ambeed
Warning

A528382-1G A528382-250MG A528382-5G
77265-990EA 55.62 USD
77265-990 77265-992 77265-994
5,9-Dibromo-7H-benzo[c]fluoren-7-one 98+%
5,9-Dibromo-7H-benzo[c]fluoren-7-one

Precaución: for Research Use Only

Formula: C₁₇H₈Br₂O
MW: 388.05 g/mol
MDL Number: MFCD30607242
CAS Number: 1637660-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR