(2S,5R)-Benzyl 5-((benzyloxy)amino)piperidine-2-carboxylate oxalate 97%

Supplier: Ambeed
Warning

A528259-100MG A528259-1G A528259-250MG
77253-562EA 5 USD
77253-562 77253-564 77253-566
(2S,5R)-Benzyl 5-((benzyloxy)amino)piperidine-2-carboxylate oxalate 97%
(2S,5R)-Benzyl 5-((benzyloxy)amino)piperidine-2-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₇
MDL Number: MFCD29472311
CAS Number: 1171080-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR