4,5-Dimethoxy-3-iodobenzaldehyde 95%

Supplier: Ambeed
Warning

A528190-1G A528190-5G
77279-836EA 18 USD
77279-836 77279-838
4,5-Dimethoxy-3-iodobenzaldehyde 95%
4,5-Dimethoxy-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉IO₃
MDL Number: MFCD00674091
CAS Number: 32024-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR