Importazole 98%

Supplier: Ambeed
Warning

A528010-10MG A528010-1MG A528010-5MG A528010-100MG
77265-976EA 26 USD
77265-976 77265-978 77265-980 77749-100
Importazole 98%
Importazole

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄
MW: 318.42 g/mol
MDL Number: MFCD04077630
CAS Number: 662163-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR