Phenylsulfonylacetone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(Phenylsulphonyl)acetone, (2-Propanoylsulphonyl)benzol, 1-(Phenylsulphonyl)propan-2-one, Benzenesulphonylacetone
A527166-1G
A527166-250MG
A527166-25G
A527166-5G
77253-548EA
28.84
USD
77253-548
77253-550
77253-552
77253-554
Phenylsulfonylacetone 98%
Phenylsulphonylacetone
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃S MW: 198.24 g/mol |
MDL Number:
MFCD00047794 CAS Number: 5000-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...