4-Hydroxybutanone 95%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

A527158-10G A527158-25G A527158-5G
77292-300EA 8.24 USD
77292-300 77292-302 77292-304
4-Hydroxybutanone 95%
4-Hydroxybutanone

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD00059005
CAS Number: 590-90-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR