2-Propyl-1-heptanol 96%

Supplier: Ambeed
Warning

A527050-100G A527050-1G A527050-25G A527050-5G
77279-828EA 267.01 USD
77279-828 77279-830 77279-832 77279-834
2-Propyl-1-heptanol 96%
2-Propyl-1-heptanol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O
MDL Number: MFCD00046768
CAS Number: 10042-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR