2,7-Diphenyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A526813-10G A526813-1G A526813-5G
77241-756EA 168 USD
77241-756 77241-758 77241-760
2,7-Diphenyl-9H-carbazole 98%
2,7-Diphenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₇N
MDL Number: MFCD30478534
CAS Number: 42448-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR