(S)-Methyl 3-(7-bromo-2-oxo-5-(pyridin-2-yl)-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanoate 97%

Supplier: Ambeed
Warning

A526680-1G A526680-25G A526680-5G
77253-542EA 33.99 USD
77253-542 77253-544 77253-546
(S)-Methyl 3-(7-bromo-2-oxo-5-(pyridin-2-yl)-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanoate 97%
(S)-Methyl 3-(7-bromo-2-oxo-5-(pyridin-2-yl)-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₆BrN₃O₃
MW: 402.24 g/mol
MDL Number: MFCD28978567
CAS Number: 308242-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR