2,3-Diphenylpyrazine 97%

Supplier: Ambeed
Danger

A526606-1G A526606-25G
77253-538EA 31 USD
77253-538 77253-540
2,3-Diphenylpyrazine 97%
2,3-Diphenylpyrazine

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.29 g/mol
MDL Number: MFCD00053000
CAS Number: 1588-89-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR