3',4'-Difluoropropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Difluorophenyl)propan-1-one

A526420-100G A526420-10G A526420-1G A526420-25G A526420-5G
77265-956EA 380.07 USD
77265-956 77265-958 77265-960 77265-962 77749-099
3',4'-Difluoropropiophenone 98%
3',4'-Difluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00015505
CAS Number: 23384-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR