2-Bromo-1-(3-fluoro-4-nitrophenyl)ethanone 97%

Supplier: Ambeed
Danger

A526112-100MG A526112-1G A526112-250MG
77241-744EA 171 USD
77241-744 77241-746 77241-748
2-Bromo-1-(3-fluoro-4-nitrophenyl)ethanone 97%
2-Bromo-1-(3-fluoro-4-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₃
MDL Number: MFCD23711756
CAS Number: 1010384-51-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR