4-(4-Butylphenylazo)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Butyl-4'-hydroxyazobenzene

A525857-1G A525857-25G A525857-5G
77292-290EA 13 USD
77292-290 77292-292 77292-294
4-(4-Butylphenylazo)phenol 98%
4-(4-Butylphenylazo)phenol

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₂O
MW: 254.33 g/mol
MDL Number: MFCD00430795
CAS Number: 2496-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR