4-Fluoro-3-(trifluoromethyl)anisole 97%

Supplier: Ambeed
Warning

A525717-1G A525717-25G
77241-740EA 41.99 USD
77241-740 77241-742
4-Fluoro-3-(trifluoromethyl)anisole 97%
4-Fluoro-3-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD01631405
CAS Number: 127271-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR