3-Bromo-5-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-fluorophenylboronic acid, (3-Bromo-5-fluorophenyl)boronic acid

A525558-100G A525558-1G A525558-25G A525558-5G
77279-816EA 536.63 USD
77279-816 77279-818 77279-820 77279-822
3-Bromo-5-fluorobenzeneboronic acid 98%
3-Bromo-5-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD06411356
CAS Number: 849062-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR