(2-Chloro-4-nitrophenyl)methanol 95%

Supplier: Ambeed
Warning

A525493-1G A525493-250MG A525493-25G A525493-5G
77265-948EA 19 USD
77265-948 77265-950 77265-952 77265-954
(2-Chloro-4-nitrophenyl)methanol 95%
(2-Chloro-4-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD09998267
CAS Number: 52301-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR