(±)-4-Fluoromandelic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoromandelic Acid, (±)-4-Fluorophenylglycolic acid

A525092-1G A525092-5G
77241-738EA 6 USD
77241-738 77747-072
(±)-4-Fluoromandelic acid 97%
(±)-4-Fluoromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD00044402
CAS Number: 395-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR