Isophorone ≥99%

Supplier: Ambeed
Warning

Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone

A524790-500G A524790-25G A524790-1000G A524790-100G
77279-814EA 12.36 USD
77279-814 77551-578 77617-236 77617-238
Isophorone ≥99%
Isophorone

Precaución: for Research Use Only

Formula: C₉H₁₄O
MW: 138.21 g/mol
MDL Number: MFCD00001584
CAS Number: 78-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR