4-Methoxythioanisole 97%

Supplier: Ambeed
Warning

A524649-1G A524649-25G A524649-5G A524649-100G
77241-726EA 6 USD
77241-726 77241-728 77241-730 77747-071
4-Methoxythioanisole 97%
4-Methoxythioanisole

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MDL Number: MFCD00010587
CAS Number: 1879-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR