2-Bromo-6-benzyloxypyridine 98%

Supplier: Ambeed
Warning

A524567-1G A524567-25G A524567-5G
77265-940EA 18 USD
77265-940 77265-942 77265-944
2-Bromo-6-benzyloxypyridine 98%
2-Bromo-6-benzyloxypyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MDL Number: MFCD00234782
CAS Number: 117068-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR